Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCAGCTGACCTGTAATTTTCTG[A/G]CCTAGTGTCTGCACTGCTTGGGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610714 MIM: 600960 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PKN3 - protein kinase N3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SET - SET nuclear proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122821.1 | Intron | NP_001116293.1 | ||||
NM_001248000.1 | Intron | NP_001234929.1 | ||||
NM_001248001.1 | Intron | NP_001234930.1 | ||||
NM_003011.3 | Intron | NP_003002.2 | ||||
XM_017015013.1 | Intron | XP_016870502.1 | ||||
XM_017015014.1 | Intron | XP_016870503.1 | ||||
XM_017015015.1 | Intron | XP_016870504.1 | ||||
XM_017015016.1 | Intron | XP_016870505.1 |