Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTTACCCAGCAGGAGAAGGACAGC[C/T]TGGTCTACCAGTATCTGCAGAAGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609682 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCLRE1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DCLRE1A - DNA cross-link repair 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271816.1 | 350 | Intron | NP_001258745.1 | |||
NM_014881.4 | 350 | Intron | NP_055696.3 | |||
XM_006718090.1 | 350 | Intron | XP_006718153.1 | |||
XM_011540429.1 | 350 | Intron | XP_011538731.1 |
NHLRC2 - NHL repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198514.3 | 350 | Silent Mutation | CTG,TTG | L,L 37 | NP_940916.2 | |
XM_011539769.2 | 350 | Silent Mutation | CTG,TTG | L,L 37 | XP_011538071.1 |
Set Membership: |
HapMap |