Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCACAGAGAGAGTGTGGTTAAG[C/T]GCATAGGCGTCGCTATCCTCTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601206 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4491 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4491 - microRNA 4491 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POU2AF1 - POU class 2 associating factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006235.2 | 2694 | Silent Mutation | GCA,GCG | A,A 246 | NP_006226.2 | |
XM_005271593.1 | 2694 | Silent Mutation | GCA,GCG | A,A 293 | XP_005271650.1 | |
XM_005271594.3 | 2694 | Silent Mutation | GCA,GCG | A,A 293 | XP_005271651.1 | |
XM_006718859.1 | 2694 | Silent Mutation | GCA,GCG | A,A 266 | XP_006718922.1 | |
XM_006718860.3 | 2694 | UTR 3 | XP_006718923.1 | |||
XM_017017932.1 | 2694 | UTR 3 | XP_016873421.1 |