Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACAGCTGGAGCAACGACTCGGAC[A/T]CCGTCCGCTCTCTGAAGCCTTGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188410 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD1D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD1D - CD1d molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319145.1 | 368 | Missense Mutation | ACC,TCC | T,S 64 | NP_001306074.1 | |
NM_001766.3 | 368 | Missense Mutation | ACC,TCC | T,S 64 | NP_001757.1 | |
XM_005245583.1 | 368 | UTR 5 | XP_005245640.1 | |||
XM_006711621.1 | 368 | UTR 5 | XP_006711684.1 | |||
XM_011510125.1 | 368 | Missense Mutation | ACC,TCC | T,S 64 | XP_011508427.1 | |
XM_011510127.1 | 368 | UTR 5 | XP_011508429.1 |