Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAATTCTACCAGCCCCAGAGGAG[G/T]TGCCATGTAAATTCCAACTAATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601129 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP2C8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYP2C8 - cytochrome P450 family 2 subfamily C member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000770.3 | Intron | NP_000761.3 | ||||
NM_001198853.1 | Intron | NP_001185782.1 | ||||
NM_001198854.1 | Intron | NP_001185783.1 | ||||
NM_001198855.1 | Intron | NP_001185784.1 |
Set Membership: |
HapMap |