Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGTTCAGTGTTTCAGTAATAGA[C/T]ATCTGTGCTAGATACTGTGGGGGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602534 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRC57 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRRC57 - leucine rich repeat containing 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153260.2 | 1132 | Intron | NP_694992.2 | |||
XM_011521423.2 | 1132 | Intron | XP_011519725.1 | |||
XM_011521424.2 | 1132 | Intron | XP_011519726.1 |
SNAP23 - synaptosome associated protein 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003825.3 | 1132 | Intron | NP_003816.2 | |||
NM_130798.2 | 1132 | Intron | NP_570710.1 | |||
XM_006720725.2 | 1132 | Intron | XP_006720788.1 | |||
XM_017022692.1 | 1132 | Missense Mutation | CAT,TAT | H,Y 222 | XP_016878181.1 | |
XM_017022693.1 | 1132 | Intron | XP_016878182.1 | |||
XM_017022694.1 | 1132 | Intron | XP_016878183.1 |