Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGGGAGAGATGAGGGTTTCAGG[G/T]AGAAAGGGGGCACCCTGCTACCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137168 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GGT5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GGT5 - gamma-glutamyltransferase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099781.2 | Intron | NP_001093251.1 | ||||
NM_001099782.2 | Intron | NP_001093252.1 | ||||
NM_001302464.1 | Intron | NP_001289393.1 | ||||
NM_001302465.1 | Intron | NP_001289394.1 | ||||
NM_004121.3 | Intron | NP_004112.2 | ||||
XM_005261557.2 | Intron | XP_005261614.1 | ||||
XM_005261558.2 | Intron | XP_005261615.1 | ||||
XM_011530133.1 | Intron | XP_011528435.1 | ||||
XM_011530134.1 | Intron | XP_011528436.1 | ||||
XM_011530135.1 | Intron | XP_011528437.1 | ||||
XM_011530136.1 | Intron | XP_011528438.1 | ||||
XM_011530137.2 | Intron | XP_011528439.1 | ||||
XM_017028768.1 | Intron | XP_016884257.1 | ||||
XM_017028769.1 | Intron | XP_016884258.1 |