Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCAAAATGTAGCCTTTATTTACT[C/T]AAACATTTATTTGCTTCTAGGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611326 MIM: 604122 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCPG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCPG1 - cell cycle progression 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204450.1 | 7047 | UTR 3 | NP_001191379.1 | |||
NM_001204451.1 | 7047 | UTR 3 | NP_001191380.1 | |||
NM_004748.4 | 7047 | UTR 3 | NP_004739.3 | |||
NM_020739.3 | 7047 | UTR 3 | NP_065790.2 |
DYX1C1-CCPG1 - DYX1C1-CCPG1 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIGB - phosphatidylinositol glycan anchor biosynthesis class B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004855.4 | 7047 | Intron | NP_004846.4 | |||
XM_005254795.4 | 7047 | Intron | XP_005254852.1 | |||
XM_011522235.2 | 7047 | Intron | XP_011520537.1 | |||
XM_011522236.2 | 7047 | Intron | XP_011520538.1 | |||
XM_011522237.2 | 7047 | Intron | XP_011520539.1 | |||
XM_017022730.1 | 7047 | Intron | XP_016878219.1 | |||
XM_017022731.1 | 7047 | Intron | XP_016878220.1 | |||
XM_017022732.1 | 7047 | Intron | XP_016878221.1 |