Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGCAGTGCCTGCCTAGTGGTCAC[A/G]GTGGATGAACATGGTCACTGGTGAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 605320 | |||||||||||||||||||||||
Literature Links: |
PFKFB4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PFKFB4 - 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317134.1 | 1504 | UTR 3 | NP_001304063.1 | |||
NM_001317135.1 | 1504 | UTR 3 | NP_001304064.1 | |||
NM_001317136.1 | 1504 | UTR 3 | NP_001304065.1 | |||
NM_001317137.1 | 1504 | UTR 3 | NP_001304066.1 | |||
NM_001317138.1 | 1504 | UTR 3 | NP_001304067.1 | |||
NM_004567.3 | 1504 | UTR 3 | NP_004558.1 | |||
XM_011533829.2 | 1504 | UTR 3 | XP_011532131.1 | |||
XM_017006614.1 | 1504 | UTR 3 | XP_016862103.1 | |||
XM_017006615.1 | 1504 | UTR 3 | XP_016862104.1 | |||
XM_017006616.1 | 1504 | UTR 3 | XP_016862105.1 | |||
XM_017006617.1 | 1504 | UTR 3 | XP_016862106.1 |