Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGGGGCTTCCAGGGATGTGGGT[A/G]CCGTTGAACTTGACGTCAATCAGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300017 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLNA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FLNA - filamin A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110556.1 | 7270 | Silent Mutation | GGC,GGT | G,G 2408 | NP_001104026.1 | |
NM_001456.3 | 7270 | Silent Mutation | GGC,GGT | G,G 2400 | NP_001447.2 | |
XM_011531127.1 | 7270 | Silent Mutation | GGC,GGT | G,G 2376 | XP_011529429.1 | |
XM_011531128.1 | 7270 | Silent Mutation | GGC,GGT | G,G 2368 | XP_011529430.1 | |
XM_011531129.1 | 7270 | Silent Mutation | GGC,GGT | G,G 2350 | XP_011529431.1 | |
XM_011531130.1 | 7270 | Silent Mutation | GGC,GGT | G,G 2342 | XP_011529432.1 | |
XM_011531131.1 | 7270 | Silent Mutation | GGC,GGT | G,G 2341 | XP_011529433.1 |
LOC105373386 - uncharacterized LOC105373386 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |