Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTACCCCTACTTGTGAAGGTGCA[C/T]GAGGAAGATGAAGCTGGGATCGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603659 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLP2R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLP2R - glucagon like peptide 2 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004246.2 | 549 | UTR 5 | NP_004237.1 | |||
XM_005256861.3 | 549 | Intron | XP_005256918.1 | |||
XM_011524077.2 | 549 | UTR 5 | XP_011522379.1 | |||
XM_017025339.1 | 549 | UTR 5 | XP_016880828.1 | |||
XM_017025340.1 | 549 | UTR 5 | XP_016880829.1 | |||
XM_017025341.1 | 549 | UTR 5 | XP_016880830.1 |
GSG1L2 - GSG1 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |