Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCACTATCCGGGTGGGGGCCCCA[C/T]GTGTTCTCAGTGTCTCAACAACATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607254 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APOL4 - apolipoprotein L4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030643.4 | 1247 | Missense Mutation | CAT,CGT | H,R 275 | NP_085146.2 | |
NM_145660.2 | 1247 | Missense Mutation | CAT,CGT | H,R 278 | NP_663693.1 |