Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607874 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF444 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZNF444 - zinc finger protein 444 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001253792.1 | Intron | NP_001240721.1 | ||||
NM_018337.3 | Intron | NP_060807.2 | ||||
XM_005259034.2 | Intron | XP_005259091.1 | ||||
XM_005259035.1 | Intron | XP_005259092.1 | ||||
XM_005259036.1 | Intron | XP_005259093.1 | ||||
XM_011527068.1 | Intron | XP_011525370.1 | ||||
XM_011527069.1 | Intron | XP_011525371.1 | ||||
XM_011527070.1 | Intron | XP_011525372.1 | ||||
XM_011527071.1 | Intron | XP_011525373.1 | ||||
XM_011527073.1 | Intron | XP_011525375.1 | ||||
XM_017026932.1 | Intron | XP_016882421.1 | ||||
XM_017026933.1 | Intron | XP_016882422.1 |