Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCCCAAAGAATTACATGTGTCCCT[C/T]GGGAACTGTAGGGCAGAGCTACCTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
30 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120326 MIM: 610033 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL16A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COL16A1 - collagen type XVI alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001856.3 | Intron | NP_001847.3 | ||||
XM_005270481.1 | Intron | XP_005270538.1 | ||||
XM_011540722.1 | Intron | XP_011539024.1 | ||||
XM_011540723.1 | Intron | XP_011539025.1 | ||||
XM_011540724.1 | Intron | XP_011539026.1 | ||||
XM_011540726.2 | Intron | XP_011539028.1 | ||||
XM_011540727.2 | Intron | XP_011539029.1 | ||||
XM_011540728.2 | Intron | XP_011539030.1 | ||||
XM_011540729.1 | Intron | XP_011539031.1 | ||||
XM_011540730.1 | Intron | XP_011539032.1 | ||||
XM_017000338.1 | Intron | XP_016855827.1 | ||||
XM_017000339.1 | Intron | XP_016855828.1 | ||||
XM_017000340.1 | Intron | XP_016855829.1 | ||||
XM_017000341.1 | Intron | XP_016855830.1 |
PEF1 - penta-EF-hand domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |