Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCGGGCGCTCCCGCAGGGGTT[C/T]CTCCAAGATGGCGGCGCAGAGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602615 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TAB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TAB1 - TGF-beta activated kinase 1/MAP3K7 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006116.2 | 42 | UTR 5 | NP_006107.1 | |||
NM_153497.2 | 42 | UTR 5 | NP_705717.1 |