Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGGGGTGGAGGTCAGGCCAGCC[G/T]ACCGGCCGGGGGCGTGGCTTTAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 100710 MIM: 601514 MIM: 613936 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRNB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRNB1 - cholinergic receptor nicotinic beta 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000747.2 | Intron | NP_000738.2 |
FGF11 - fibroblast growth factor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303460.1 | Intron | NP_001290389.1 | ||||
NM_004112.3 | Intron | NP_004103.1 |
TMEM102 - transmembrane protein 102 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |