Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACAGGAGCTGGGCTAGAAGGGT[C/G]AGGGTGGGGGAGTAGGGGCAGGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600187 MIM: 601935 MIM: 615865 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF5A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF5A - eukaryotic translation initiation factor 5A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GPS2 - G protein pathway suppressor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEURL4 - neuralized E3 ubiquitin protein ligase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005408.1 | Intron | NP_001005408.1 | ||||
NM_032442.2 | Intron | NP_115818.2 |