Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCATGGTCTCCATCATAGAACTCA[C/T]TGGGTGCCTCTGGCCTGCAAAGCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602121 MIM: 605166 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DIAPH1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DIAPH1 - diaphanous related formin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HDAC3 - histone deacetylase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003883.3 | 1298 | Missense Mutation | AAT,AGT | N,S 411 | NP_003874.2 | |
XM_011537697.2 | 1298 | Missense Mutation | AAT,AGT | N,S 224 | XP_011535999.1 |