Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGCATCAGCTCCCTAACGGGTAA[A/G]GAGAGCAGCACAGCACGCCTACCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616569 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSAD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSAD - cysteine sulfinic acid decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244705.1 | Intron | NP_001231634.1 | ||||
NM_001244706.1 | Intron | NP_001231635.1 | ||||
NM_015989.4 | Intron | NP_057073.4 | ||||
XM_011538442.2 | Intron | XP_011536744.2 | ||||
XM_011538446.2 | Intron | XP_011536748.1 | ||||
XM_011538449.1 | Intron | XP_011536751.1 | ||||
XM_011538451.1 | Intron | XP_011536753.1 | ||||
XM_017019416.1 | Intron | XP_016874905.1 | ||||
XM_017019417.1 | Intron | XP_016874906.1 | ||||
XM_017019418.1 | Intron | XP_016874907.1 | ||||
XM_017019419.1 | Intron | XP_016874908.1 |