Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGTCAGCAAGGAGCAGAATCCAAC[C/G]ATCGGGGCTTTTCAAAGCCCCGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB9 - ankyrin repeat and SOCS box containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031739.2 | Intron | NP_001026909.1 | ||||
NM_001168530.1 | Intron | NP_001162002.1 | ||||
NM_001168531.1 | Intron | NP_001162003.1 | ||||
NM_024087.2 | Intron | NP_076992.1 | ||||
XM_005274446.1 | Intron | XP_005274503.1 | ||||
XM_011545458.1 | Intron | XP_011543760.1 | ||||
XM_011545460.1 | Intron | XP_011543762.1 | ||||
XM_017029283.1 | Intron | XP_016884772.1 | ||||
XM_017029284.1 | Intron | XP_016884773.1 | ||||
XM_017029285.1 | Intron | XP_016884774.1 | ||||
XM_017029286.1 | Intron | XP_016884775.1 |