Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACTCCATTTAAATACATGCAACTA[C/T]GTGCATATTTTGTAGCAGTCAGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H7A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC3H7A - zinc finger CCCH-type containing 7A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014153.3 | Intron | NP_054872.2 | ||||
XM_006720877.2 | Intron | XP_006720940.1 | ||||
XM_011522463.1 | Intron | XP_011520765.1 | ||||
XM_011522464.2 | Intron | XP_011520766.1 | ||||
XM_017023174.1 | Intron | XP_016878663.1 | ||||
XM_017023175.1 | Intron | XP_016878664.1 |