Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300674 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOSPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MOSPD1 - motile sperm domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001306188.1 | 1377 | UTR 3 | NP_001293117.1 | |||
NM_019556.2 | 1377 | UTR 3 | NP_062456.1 | |||
XM_011531366.2 | 1377 | UTR 3 | XP_011529668.1 | |||
XM_017029653.1 | 1377 | UTR 3 | XP_016885142.1 | |||
XM_017029654.1 | 1377 | UTR 3 | XP_016885143.1 |