Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGCTATGTTGCTCTATCTTCCAC[A/G]TCCAAAAACAGTCCTATGTAGCTTC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 615020 | |||||||||||||||||||||||
Literature Links: |
ELP6 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ELP6 - elongator acetyltransferase complex subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031703.2 | 1235 | UTR 3 | NP_001026873.2 | |||
XM_005265241.3 | 1235 | UTR 3 | XP_005265298.1 | |||
XM_011533851.2 | 1235 | Intron | XP_011532153.1 | |||
XM_011533852.2 | 1235 | Intron | XP_011532154.1 | |||
XM_011533853.2 | 1235 | Intron | XP_011532155.1 | |||
XM_011533854.2 | 1235 | Intron | XP_011532156.1 | |||
XM_011533856.2 | 1235 | Intron | XP_011532158.1 | |||
XM_011533857.2 | 1235 | Intron | XP_011532159.1 | |||
XM_017006669.1 | 1235 | UTR 3 | XP_016862158.1 | |||
XM_017006670.1 | 1235 | UTR 3 | XP_016862159.1 |