Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGTTTCTGCGCCGGACCAGTCG[A/C]GGACTCTGGACAGTAGAGGCCCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605363 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GAD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GAD1 - glutamate decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000817.2 | 382 | UTR 5 | NP_000808.2 | |||
NM_013445.3 | 382 | UTR 5 | NP_038473.2 | |||
XM_005246444.2 | 382 | UTR 5 | XP_005246501.1 | |||
XM_011510922.1 | 382 | UTR 5 | XP_011509224.1 | |||
XM_017003756.1 | 382 | UTR 5 | XP_016859245.1 | |||
XM_017003757.1 | 382 | UTR 5 | XP_016859246.1 | |||
XM_017003758.1 | 382 | UTR 5 | XP_016859247.1 |