Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAATGTACCAGGGTGGCTCGGGTT[C/T]GCGTCTAAAGGTTTCTTCCTCCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605537 MIM: 604835 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATF6 - activating transcription factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007348.3 | Intron | NP_031374.2 | ||||
XM_006711224.1 | Intron | XP_006711287.1 | ||||
XM_011509308.1 | Intron | XP_011507610.1 | ||||
XM_011509309.1 | Intron | XP_011507611.1 | ||||
XM_011509310.2 | Intron | XP_011507612.1 |
DUSP12 - dual specificity phosphatase 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |