Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCAATCCGAGTGTTGATGGGGGT[C/T]GAGCTGCTCTGACCACAGTGTTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606704 | ||||||||||||||||||||
Literature Links: |
GPR75 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR75 - G protein-coupled receptor 75 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006794.3 | 1731 | Silent Mutation | NP_006785.1 |
GPR75-ASB3 - GPR75-ASB3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164165.1 | 1731 | Intron | NP_001157637.1 |
MIR3682 - microRNA 3682 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |