Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGGGGGCAGCGCGGGCGGCTCC[A/G]ACGTGACGGGGATGGAGAGGCTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607679 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOCK4 - dedicator of cytokinesis 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014705.3 | 6578 | Missense Mutation | TCG,TTG | S,L 1926 | NP_055520.3 | |
XM_006716188.2 | 6578 | Missense Mutation | TCG,TTG | S,L 1935 | XP_006716251.1 | |
XM_006716189.2 | 6578 | Missense Mutation | TCG,TTG | S,L 1897 | XP_006716252.1 | |
XM_017012819.1 | 6578 | Missense Mutation | TCG,TTG | S,L 1966 | XP_016868308.1 | |
XM_017012820.1 | 6578 | Missense Mutation | TCG,TTG | S,L 1957 | XP_016868309.1 | |
XM_017012821.1 | 6578 | Missense Mutation | TCG,TTG | S,L 1948 | XP_016868310.1 | |
XM_017012822.1 | 6578 | Missense Mutation | TCG,TTG | S,L 1946 | XP_016868311.1 | |
XM_017012823.1 | 6578 | Missense Mutation | TCG,TTG | S,L 1928 | XP_016868312.1 | |
XM_017012824.1 | 6578 | Missense Mutation | TCG,TTG | S,L 1919 | XP_016868313.1 | |
XM_017012825.1 | 6578 | Missense Mutation | TCG,TTG | S,L 1910 | XP_016868314.1 |