Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGATAAATGAGAGACAGTGAAG[C/T]GCTCTAATGGGCTAGCAACACCACA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610584 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf131 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
C1orf131 - chromosome 1 open reading frame 131 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300830.1 | Intron | NP_001287759.1 | ||||
NM_152379.3 | Intron | NP_689592.2 |
TRIM67 - tripartite motif containing 67 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |