Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616978 MIM: 602000 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHCHD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHCHD5 - coiled-coil-helix-coiled-coil-helix domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304353.1 | 869 | UTR 5 | NP_001291282.1 | |||
NM_001304354.1 | 869 | Intron | NP_001291283.1 | |||
NM_032309.3 | 869 | UTR 5 | NP_115685.1 | |||
XM_005263824.3 | 869 | UTR 5 | XP_005263881.1 |
POLR1B - RNA polymerase I subunit B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |