Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 229000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLKB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLKB1 - kallikrein B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000892.4 | Intron | NP_000883.2 | ||||
NM_001318394.1 | Intron | NP_001305323.1 | ||||
NM_001318396.1 | Intron | NP_001305325.1 | ||||
XM_011531930.2 | Intron | XP_011530232.1 | ||||
XM_017008181.1 | Intron | XP_016863670.1 | ||||
XM_017008182.1 | Intron | XP_016863671.1 | ||||
XM_017008183.1 | Intron | XP_016863672.1 | ||||
XM_017008184.1 | Intron | XP_016863673.1 |