Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGCCCACCATGACCCCTCCCTC[A/G]GGGGGCACCCCCACAGTCACGGTAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608508 MIM: 604628 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYBA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CYBA - cytochrome b-245 alpha chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL17C - interleukin 17C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013278.3 | 122 | Missense Mutation | AGG,GGG | R,G 25 | NP_037410.1 |
ZC3H18 - zinc finger CCCH-type containing 18 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |