Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGAGCTGACATTCTTGGAAGTAGC[C/T]CCGACTGCTTGGACAAGATCATTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614503 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHDC8A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KLHDC8A - kelch domain containing 8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271863.1 | 1679 | UTR 3 | NP_001258792.1 | |||
NM_001271864.1 | 1679 | UTR 3 | NP_001258793.1 | |||
NM_001271865.1 | 1679 | UTR 3 | NP_001258794.1 | |||
NM_018203.2 | 1679 | UTR 3 | NP_060673.1 |