Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGCCTCTTACAGTTTTCTGAGA[C/T]TGGTAGGATTTGGAGCTATAGAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614503 | ||||||||||||||||||||
Literature Links: |
KLHDC8A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KLHDC8A - kelch domain containing 8A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271863.1 | 1772 | UTR 3 | NP_001258792.1 | |||
NM_001271864.1 | 1772 | UTR 3 | NP_001258793.1 | |||
NM_001271865.1 | 1772 | UTR 3 | NP_001258794.1 | |||
NM_018203.2 | 1772 | UTR 3 | NP_060673.1 |