Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAAAAGATTAGCAGTAATATTA[A/G]GAGAAATTTTTGGGCTTGTCAGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616014 MIM: 607652 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF25 - ring finger protein 25 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK36 - serine/threonine kinase 36 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243313.1 | Intron | NP_001230242.1 | ||||
NM_015690.4 | Intron | NP_056505.2 | ||||
XM_005246464.1 | Intron | XP_005246521.1 | ||||
XM_011510959.1 | Intron | XP_011509261.1 | ||||
XM_011510960.1 | Intron | XP_011509262.1 | ||||
XM_011510961.1 | Intron | XP_011509263.1 | ||||
XM_017003804.1 | Intron | XP_016859293.1 | ||||
XM_017003805.1 | Intron | XP_016859294.1 |