Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606877 MIM: 613331 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANP32C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANP32C - acidic nuclear phosphoprotein 32 family member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012403.1 | Intron | NP_036535.1 |
MARCH1 - membrane associated ring-CH-type finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166373.1 | Intron | NP_001159845.1 | ||||
NM_017923.3 | Intron | NP_060393.1 | ||||
XM_011532055.2 | Intron | XP_011530357.1 | ||||
XM_017008334.1 | Intron | XP_016863823.1 | ||||
XM_017008335.1 | Intron | XP_016863824.1 | ||||
XM_017008336.1 | Intron | XP_016863825.1 |
Set Membership: |
HapMap |