Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCCCGGCTTCCTGGGGACACAGG[G/T]TTGGCACCATGGGGCCCACCAGCGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601974 | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928370 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LOC101928370 - uncharacterized LOC101928370 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
S1PR1 - sphingosine-1-phosphate receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320730.1 | 398 | UTR 5 | NP_001307659.1 | |||
NM_001400.4 | 398 | UTR 5 | NP_001391.2 |
Set Membership: |
HapMap JSNP Validated |