Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTATTTAATGCTACCAAGAGAAGA[A/G]TATTTTTCAGAAATATAAAGATTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
37 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604003 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLCA2 - chloride channel accessory 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006536.5 | 400 | Missense Mutation | ATA,GTA | I,V 80 | NP_006527.1 | |
XM_011542448.2 | 400 | Missense Mutation | ATA,GTA | I,V 80 | XP_011540750.1 |
Set Membership: |
HapMap |