Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606715 MIM: 615495 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASIC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASIC4 - acid sensing ion channel subunit family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182847.2 | 669 | Silent Mutation | GCC,GCG | A,A 165 | NP_878267.2 | |
XM_005246671.2 | 669 | Silent Mutation | GCC,GCG | A,A 165 | XP_005246728.2 | |
XM_017004439.1 | 669 | Intron | XP_016859928.1 |
GMPPA - GDP-mannose pyrophosphorylase A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |