Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTGGAAATGGATCTGGAACATT[C/T]CAGATCTATTCCTGGATCCCTTTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608359 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105373057 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105373057 - DNA-directed RNA polymerase II subunit RPB1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SULT4A1 - sulfotransferase family 4A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014351.3 | Intron | NP_055166.1 | ||||
XM_011530120.2 | Intron | XP_011528422.1 | ||||
XM_011530121.1 | Intron | XP_011528423.1 |