Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTTTAGCTTATCCCACAGCGTCT[C/T]ATGTCTGCCAGCAATAGCAATGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614843 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ODAM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ODAM - odontogenic, ameloblast asssociated | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017855.3 | 86 | Silent Mutation | CTC,CTT | L,L 23 | NP_060325.3 | |
XM_005265698.3 | 86 | Silent Mutation | CTC,CTT | L,L 40 | XP_005265755.1 | |
XM_006714245.3 | 86 | Silent Mutation | CTC,CTT | L,L 40 | XP_006714308.1 | |
XM_017008331.1 | 86 | Intron | XP_016863820.1 |