Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCACTGCCTGGCCCCCATGCCCAC[C/T]GCAGAGGAGCACTCACCCTTGGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120260 MIM: 606480 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL9A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL9A2 - collagen type IX alpha 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001852.3 | Intron | NP_001843.1 | ||||
XM_006710365.3 | Intron | XP_006710428.1 | ||||
XM_011540715.2 | Intron | XP_011539017.1 | ||||
XM_011540716.2 | Intron | XP_011539018.1 | ||||
XM_011540717.2 | Intron | XP_011539019.1 | ||||
XM_017000332.1 | Intron | XP_016855821.1 | ||||
XM_017000333.1 | Intron | XP_016855822.1 |
ZMPSTE24 - zinc metallopeptidase STE24 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |