Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCCAAAGATAACATGGGTCTCA[A/T]TTCAAGGACTGACCCCTAATACAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 137140 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRA2 - gamma-aminobutyric acid type A receptor alpha2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005248080.3 | 1523 | UTR 3 | XP_005248137.1 | |||
XM_011513675.1 | 1523 | UTR 3 | XP_011511977.1 | |||
XM_011513676.1 | 1523 | UTR 3 | XP_011511978.1 | |||
XM_017007982.1 | 1523 | UTR 3 | XP_016863471.1 | |||
XM_017007983.1 | 1523 | UTR 3 | XP_016863472.1 |