Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCTGGCCACTTTGCCTGAATTC[A/G]TGGTTTATAAACCTCAGATGGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608379 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCRL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCRL2 - C-C motif chemokine receptor like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130910.1 | 787 | Missense Mutation | ATG,GTG | M,V 180 | NP_001124382.1 | |
NM_003965.4 | 787 | Missense Mutation | ATG,GTG | M,V 168 | NP_003956.2 | |
XM_011534208.1 | 787 | Missense Mutation | ATG,GTG | M,V 168 | XP_011532510.1 | |
XM_011534209.1 | 787 | Missense Mutation | ATG,GTG | M,V 168 | XP_011532511.1 | |
XM_017007436.1 | 787 | Missense Mutation | ATG,GTG | M,V 168 | XP_016862925.1 |
LOC102724297 - uncharacterized LOC102724297 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |