Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGTTGTTGGAGTCGAATCTGGGG[A/G]TCATTCGGGGGCCAGGCATGCAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611827 | ||||||||||||||||||||
Literature Links: |
MRPL14 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL14 - mitochondrial ribosomal protein L14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318767.1 | 367 | Missense Mutation | ACC,ATC | T,I 97 | NP_001305696.1 | |
NM_001318768.1 | 367 | Missense Mutation | ACC,ATC | T,I 97 | NP_001305697.1 | |
NM_001318769.1 | 367 | Missense Mutation | ACC,ATC | T,I 108 | NP_001305698.1 | |
NM_001318770.1 | 367 | Missense Mutation | ACC,ATC | T,I 150 | NP_001305699.1 | |
NM_001318771.1 | 367 | Missense Mutation | ACC,ATC | T,I 63 | NP_001305700.1 | |
NM_032111.3 | 367 | Missense Mutation | ACC,ATC | T,I 97 | NP_115487.2 |