Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTGCTTGGGGCTAAGGGGGATAT[A/G]GGGCTTCTCTGGGGCCAACTGACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C9orf131 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C9orf131 - chromosome 9 open reading frame 131 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040410.2 | 92 | Missense Mutation | ATA,ATG | I,M 15 | NP_001035500.1 | |
NM_001040411.2 | 92 | Intron | NP_001035501.1 | |||
NM_001040412.2 | 92 | Intron | NP_001035502.1 | |||
NM_001287391.1 | 92 | Intron | NP_001274320.1 | |||
NM_203299.3 | 92 | Missense Mutation | ATA,ATG | I,M 15 | NP_976044.2 |