Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACCTTTATCTTCTGGTGCTATC[C/T]CCTGAGCTTTCTTAAGATCAGCCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 231675 MIM: 601753 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ETFDH PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ETFDH - electron transfer flavoprotein dehydrogenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPID - peptidylprolyl isomerase D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005038.2 | 1116 | Missense Mutation | GAG,GGG | E,G 335 | NP_005029.1 |
Set Membership: |
HapMap |