Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGACCCATGTTATAAGTAACATG[A/C]GATCCACATGCAATTCCAGCACCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612826 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGPP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SGPP1 - sphingosine-1-phosphate phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030791.3 | 1150 | Silent Mutation | TCG,TCT | S,S 333 | NP_110418.1 | |
XM_017021678.1 | 1150 | UTR 3 | XP_016877167.1 |