Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATTGAAAAGGACTTCCAAACAT[A/G]TCAGGCTTCTGGTTTATGGATCAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611171 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENO4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ENO4 - enolase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242699.1 | 1925 | Intron | NP_001229628.1 | |||
XM_005269815.3 | 1925 | Intron | XP_005269872.1 | |||
XM_006717835.3 | 1925 | Intron | XP_006717898.1 | |||
XM_006717836.3 | 1925 | Intron | XP_006717899.1 | |||
XM_011539797.2 | 1925 | Intron | XP_011538099.1 | |||
XM_011539799.2 | 1925 | Intron | XP_011538101.1 | |||
XM_017016241.1 | 1925 | Intron | XP_016871730.1 |
SHTN1 - shootin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127211.2 | 1925 | UTR 3 | NP_001120683.1 | |||
NM_001258298.1 | 1925 | UTR 3 | NP_001245227.1 | |||
NM_001258299.1 | 1925 | Intron | NP_001245228.1 | |||
NM_001258300.1 | 1925 | Intron | NP_001245229.1 | |||
NM_018330.6 | 1925 | UTR 3 | NP_060800.2 | |||
XM_005270002.3 | 1925 | Intron | XP_005270059.1 | |||
XM_006717931.3 | 1925 | Intron | XP_006717994.1 | |||
XM_017016462.1 | 1925 | UTR 3 | XP_016871951.1 |
Set Membership: |
HapMap |