Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACAGCAATGTACTGAAGAACTTCA[G/T]AACCTCAATTAGCCATTTGCCATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610138 | ||||||||||||||||||||
Literature Links: |
ST6GALNAC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ST6GALNAC1 - ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289107.1 | 2185 | UTR 3 | NP_001276036.1 | |||
NM_018414.4 | 2185 | UTR 3 | NP_060884.1 |