Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGGATGGCAATGAGCCCAGGACG[A/G]TCCCCTAAGGCTCTGTGGCAAGAAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605492 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371692 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105371692 - uncharacterized LOC105371692 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRN2 - leucine rich repeat neuronal 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006338.2 | 2339 | Silent Mutation | GAC,GAT | D,D 627 | NP_006329.2 | |
NM_201630.1 | 2339 | Silent Mutation | GAC,GAT | D,D 627 | NP_963924.1 | |
XM_005244827.3 | 2339 | Silent Mutation | GAC,GAT | D,D 627 | XP_005244884.1 | |
XM_011509066.2 | 2339 | Silent Mutation | GAC,GAT | D,D 627 | XP_011507368.1 | |
XM_017000050.1 | 2339 | Silent Mutation | GAC,GAT | D,D 627 | XP_016855539.1 |
Set Membership: |
HapMap JSNP |